Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Piperazinone 98.0+%, TCI America™
SDP

Catalog No. P18671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18671G 1 g
P18675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18671G Supplier TCI America Supplier No. P18671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5625-67-2
Molecular Formula C4H9N2O
Molecular Weight (g/mol) 101.13
MDL Number MFCD01318687
InChI Key IWELDVXSEVIIGI-UHFFFAOYSA-O
Synonym 2-piperazinone, 2-oxopiperazine, piperazine-2-one, piperazinone, 2-piperadone, oxopiperazine, piperazone, keto piperazine, 3-oxopiperazine, 2-ketopiperazine
PubChem CID 231360
IUPAC Name 3-oxopiperazin-1-ium
SMILES O=C1C[NH2+]CCN1

Specifications

Melting Point 134°C
Boiling Point 164°C
Quantity 1 g
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Piperazinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.