Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Piperidin-1-yl)acetic acid, Thermo Scientific™

Catalog No. AC469600010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469600010 1 g
AC469600050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469600010 Supplier Thermo Scientific Chemicals Supplier No. 469600010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Specifications

Chemical Name or Material 2-(Piperidin-1-yl)acetic acid
Alpha Vector PIPERIDINYLACETICACID
CAS 3235-67-4
Melting Point 210°C to 216°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C7 H13 N O2
MDL Number 00461042
Quantity 1 g
Molecular Weight (g/mol) 143.19
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.