Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Piperidinecarboxamide 98.0+%, TCI America™
SDP

Catalog No. P20281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20281G Supplier TCI America Supplier No. P20281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19889-77-1
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD05663784
InChI Key XIMBESZRBTVIOD-UHFFFAOYNA-N
PubChem CID 140623
IUPAC Name piperidine-2-carboxamide
SMILES NC(=O)C1CCCCN1

Specifications

Melting Point 146°C
Color White
Quantity 1 g
Formula Weight 128.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Piperidinecarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.