Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Piperidinemethanol, 97%

Catalog No. p-4228880
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168030050 5 g Glass bottle
AC168030250 25 g Glass bottle
AC168031000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC168030050 Supplier Thermo Scientific Chemicals Supplier No. 168030050
Only null left
Add to Cart

CAS: 3433-37-2 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3433-37-2
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key PRAYXGYYVXRDDW-UHFFFAOYSA-N
Synonym 2-piperidinemethanol, 2-hydroxymethylpiperidine, 2-hydroxymethyl piperidine, 2-piperidinylmethanol, piperidin-2-yl-methanol, piperidine-2-methanol, 2-piperidylmethan-1-ol, 2-piperidine methanol, pubchem6779
PubChem CID 94263
IUPAC Name piperidin-2-ylmethanol
SMILES C1CCNC(C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 58.0°C to 64.0°C
Color White to Yellow
Boiling Point 100°C to 102°C (8.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 115.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Piperidinemethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 222-333-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.