Learn More
Medchemexpress LLC 2-piperidinone, 1-[(1S)-1-(4-fluorophenyl)ethyl]-3-[[3-methoxy-4-(4-methyl-1H-imidazol-1-yl)phenyl] | 870843-42-8 | 97.0% | 419.49 | C25H26FN3O2 | 1 ML
SDP
Catalog No. 5000215212
Encompass_Preferred
E 2012 is a small-molecule γ-secretase modulator used in preclinical research to reduce amyloid β-42 and to probe cholesterol biosynthesis via DHCR24 inhibition. Supplied for laboratory use as a concentrated solution in DMSO, it is intended for in vitro and related studies.
  • Potent γ-secretase modulator activity.
  • Reduces amyloid β-42 levels in cellular assays.
  • Inhibits 3β-hydroxysterol Δ24-reductase (DHCR24).
  • Available as a 10 mM solution in DMSO (1 mL) and as solid formats.
  • High solubility in DMSO (~119 mM) with ultrasonic assistance.
  • Manufacturer-reported purity approximately 97.0% and defined storage conditions.
Catalog No. 50-002-15212 Supplier Medchemexpress LLC Supplier No. HY1001610MM 1ML
E 2012 is a small-molecule γ-secretase modulator used in preclinical research to reduce amyloid β-42 and to probe cholesterol biosynthesis via DHCR24 inhibition. Supplied for laboratory use as a concentrated solution in DMSO, it is intended for in vitro and related studies.
  • Potent γ-secretase modulator activity.
  • Reduces amyloid β-42 levels in cellular assays.
  • Inhibits 3β-hydroxysterol Δ24-reductase (DHCR24).
  • Available as a 10 mM solution in DMSO (1 mL) and as solid formats.
  • High solubility in DMSO (~119 mM) with ultrasonic assistance.
  • Manufacturer-reported purity approximately 97.0% and defined storage conditions.