Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-piperidinone | 675-20-7 | MFCD00006037 | ≥98.0% | 99.13 g/mol | C5H9NO | 500 G
SDP

Catalog No. 5000209226
Encompass_Preferred

2-piperidinone is an endogenous metabolite and a biomarker for CYP2E1 activity, supplied as a research-grade solid with high purity and DMSO solubility.

  • Endogenous metabolite and CYP2E1 biomarker.
  • Solid powder suitable for laboratory use.
  • Purity ≥98.0% (reported by manufacturer).
  • Molecular weight 99.13 g/mol and formula C5H9NO.
  • Soluble in DMSO at approximately 100 mg/mL; may require ultrasonic assistance.
  • Store powder at -20°C for long-term stability; in solution keep at -80°C.

Catalog No. 50-002-09226 Supplier Medchemexpress LLC Supplier No. HYW042193500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-piperidinone is an endogenous metabolite and a biomarker for CYP2E1 activity, supplied as a research-grade solid with high purity and DMSO solubility.

  • Endogenous metabolite and CYP2E1 biomarker.
  • Solid powder suitable for laboratory use.
  • Purity ≥98.0% (reported by manufacturer).
  • Molecular weight 99.13 g/mol and formula C5H9NO.
  • Soluble in DMSO at approximately 100 mg/mL; may require ultrasonic assistance.
  • Store powder at -20°C for long-term stability; in solution keep at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.