Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-piperidinone | 675-20-7 | MFCD00006037 | 99.9% | 99.13 g/mol | C5H9NO | 5 G
SDP

Catalog No. 5000208827
Encompass_Preferred

2-Piperidone is an endogenous metabolite used as a biomarker for cytochrome P450 2E1 (CYP2E1) activity. It is supplied as a high-purity analytical standard for research use only.

  • Endogenous metabolite and CYP2E1 biomarker.
  • High purity: 99.9% (GC).
  • CAS number: 675-20-7.
  • Molecular formula: C5H9NO; molecular weight: 99.13 g/mol.
  • Supplied as an analytical standard for research use only.
  • Available in 5 G packaging.

Catalog No. 50-002-08827 Supplier Medchemexpress LLC Supplier No. HYW0421935G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Piperidone is an endogenous metabolite used as a biomarker for cytochrome P450 2E1 (CYP2E1) activity. It is supplied as a high-purity analytical standard for research use only.

  • Endogenous metabolite and CYP2E1 biomarker.
  • High purity: 99.9% (GC).
  • CAS number: 675-20-7.
  • Molecular formula: C5H9NO; molecular weight: 99.13 g/mol.
  • Supplied as an analytical standard for research use only.
  • Available in 5 G packaging.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.