Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-piperidone | 675-20-7 | MFCD00006037 | 99.9% | 99.13 g/mol | C5H9NO | 25 G
SDP

Catalog No. 5000209533
Encompass_Preferred

2-Piperidone is a small lactam (δ-valerolactam) used as a research reagent and a biomarker for CYP2E1 activity. It is provided as a high-purity reference standard suitable for in vitro and in vivo metabolic studies and formulation development.

  • CAS number 675-20-7.
  • Purity 99.9%.
  • Molecular formula C5H9NO.
  • Molecular weight 99.13 g/mol.
  • Solubility: DMSO and water 100 mg/mL (ultrasonic recommended); in vivo formulations ≥2.5 mg/mL in common vehicles.
  • Storage: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available pack sizes include 25 g.

Catalog No. 50-002-09533 Supplier Medchemexpress LLC Supplier No. 25GHYW042193
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Piperidone is a small lactam (δ-valerolactam) used as a research reagent and a biomarker for CYP2E1 activity. It is provided as a high-purity reference standard suitable for in vitro and in vivo metabolic studies and formulation development.

  • CAS number 675-20-7.
  • Purity 99.9%.
  • Molecular formula C5H9NO.
  • Molecular weight 99.13 g/mol.
  • Solubility: DMSO and water 100 mg/mL (ultrasonic recommended); in vivo formulations ≥2.5 mg/mL in common vehicles.
  • Storage: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available pack sizes include 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.