Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-piperidone (δ-valerolactam) | 675-20-7 | MFCD00006037 | 99.9% | 99.13 | C5H9NO | 50 G
SDP

Catalog No. 5000209227
Encompass_Preferred

2-Piperidone is a small-molecule endogenous metabolite reported as a biomarker for CYP2E1 activity. It is used in biochemical and metabolic research where measurement or modulation of metabolic enzyme activity is required.

  • chemical formula: C5H9NO
  • molecular weight: 99.13 g/mol
  • cas number: 675-20-7
  • high purity (reported): 99.9%
  • available in multiple pack sizes, including 50 G
  • soluble in common organic solvents; supplied as DMSO solution options for ready use
  • storage: pure form at -20°C (long-term) or 4°C (shorter-term)

Catalog No. 50-002-09227 Supplier Medchemexpress LLC Supplier No. HYW04219350G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Piperidone is a small-molecule endogenous metabolite reported as a biomarker for CYP2E1 activity. It is used in biochemical and metabolic research where measurement or modulation of metabolic enzyme activity is required.

  • chemical formula: C5H9NO
  • molecular weight: 99.13 g/mol
  • cas number: 675-20-7
  • high purity (reported): 99.9%
  • available in multiple pack sizes, including 50 G
  • soluble in common organic solvents; supplied as DMSO solution options for ready use
  • storage: pure form at -20°C (long-term) or 4°C (shorter-term)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.