Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-piperidone (δ-valerolactam) | 675-20-7 | MFCD00006037 | 99.9% | 99.13 g/mol | C5H9NO | 1 ML
SDP

Catalog No. 5000208499
Encompass_Preferred

2-Piperidone is an endogenous metabolite used as a biomarker for cytochrome P450 2E1 (CYP2E1) activity and has reported hepatoprotective effects in CCl4-induced hepatotoxicity models in rats.

  • Endogenous metabolite used as a biomarker for CYP2E1 activity.
  • Reported hepatoprotective effects in CCl4-induced hepatotoxicity rat models.
  • Available as a 10 mM solution (1 mL in DMSO) and as a solid in multiple pack sizes.
  • High purity (99.89%).
  • CAS number 675-20-7; molecular formula C5H9NO; molecular weight 99.13 g/mol.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • SDS and COA are provided for safe handling and quality verification.

Catalog No. 50-002-08499 Supplier Medchemexpress LLC Supplier No. HYW04219310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Piperidone is an endogenous metabolite used as a biomarker for cytochrome P450 2E1 (CYP2E1) activity and has reported hepatoprotective effects in CCl4-induced hepatotoxicity models in rats.

  • Endogenous metabolite used as a biomarker for CYP2E1 activity.
  • Reported hepatoprotective effects in CCl4-induced hepatotoxicity rat models.
  • Available as a 10 mM solution (1 mL in DMSO) and as a solid in multiple pack sizes.
  • High purity (99.89%).
  • CAS number 675-20-7; molecular formula C5H9NO; molecular weight 99.13 g/mol.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • SDS and COA are provided for safe handling and quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.