Learn More
Fisherbrand™ 2-Place Slide Mailer, PP
Catalog No. 14127544 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Assorted
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-544 Assorted
14-127-531 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-127-544
Add to Cart
Add to Cart
Snap-secure protection for every slide
  • Polypropylene protectors ideal for secure shipping and storing of slides.
  • Large, easy-to-secure snaps, provide safe storage for valuable slides.
  • Variety of colors brings some fun to the lab and enables color coding.
Specifications
Type Slide Mailer
Slide Capacity 2
Color Assorted
Dimensions (L x W x H) 3.7 x 2.7 x 0.2 in.
Material Polypropylene