Learn More
Valutek™ 2-ply Polyester Wiper , Ultrasonic seal
Catalog No. 191502025
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
12 x 12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-150-2025 12 x 12 in.
19-150-2024 9 x 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-150-2025 Supplier Valutek™ Supplier No. VT2PNWUS1212
Add to Cart
Add to Cart
This item is not returnable. View return policy
Constructed from 100% continuous filament polyester in a 2-ply double-knit, no-run, interlock pattern with ultrasonic sealed edges. This wiper is ultraclean and highly absorbent with critically low levels of particulate and extractable counts.
  • 100% continuous filament polyester knit 2-ply construction with unique pattern and knitting structure that provides the highest cleanliness
  • Ultrasonic sealed edge with reduced contamination
  • Highest possible absorbency
  • Chemically compatible with IPA and other solvents
  • Critically low levels of particulate and extractable counts Available in a variety of sizes to suit most applications
Specifications
Material Polyester
Dimensions (L x W) 12 x 12 in.
Sterility Non-sterile