Learn More
Medchemexpress LLC HY-100788 10mg , 2-PMPA CAS:173039-10-6 Purity:>98%
SDP
Catalog No. 502260382
Encompass_Preferred
HY-100788 10mg 2-PMPA CAS: 173039-10-6 2-PMPA is a potent and selective inhibitor of glutamate carboxypeptidase II (GCPII) with an IC50 of 300 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0382 Supplier Medchemexpress LLC Supplier No. HY10078810MG
HY-100788 10mg 2-PMPA CAS: 173039-10-6 2-PMPA is a potent and selective inhibitor of glutamate carboxypeptidase II (GCPII) with an IC50 of 300 pM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.