Learn More
TARGETMOL CHEMICALS INC 2-PMPA 25MG
SDP
Catalog No. 502580262
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 2-PMPA (2-(Phosphonomethyl)pentanedioic acid) is a potent and selective inhibitor of glutamate carboxypeptidase II (GCPII) (IC50=300 pM). purity: 98%
Catalog No. 50-258-0262 Supplier TARGETMOL CHEMICALS INC Supplier No. T344025MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. 2-PMPA (2-(Phosphonomethyl)pentanedioic acid) is a potent and selective inhibitor of glutamate carboxypeptidase II (GCPII) (IC50=300 pM). purity: 98%