Learn More
Alkali Scientific 2" Polycarbonate Freezer Box with 81 cell divider
SDP
Catalog No. 5000663014
Encompass_Preferred
2" Polycarbonate Freezer Box with 81 cell divider holds the most popular cryo and cold storage vials and fit into standard freezer racks. Boxes have clear lids with locator grid to facilitate sample identification. Polycarbonate boxes are designed to withstand temperature extremes of 196 to 121 . Boxes have drain holes for venting and safety. Keyed lid prevents misalignment of grid. Features: Fit all the most popular cryo and cold storage vials. Fit into standard freezer racks. Boxes have clear lids with locator grid to facilitate sample identification. Withstand temperature extremes of 196°C to 121°C. Drain holes for venting and safety. Keyed lid prevents misalignment of grid.
Catalog No. 50-006-63014 Supplier Alkali Scientific Supplier No. PCB2811
2" Polycarbonate Freezer Box with 81 cell divider holds the most popular cryo and cold storage vials and fit into standard freezer racks. Boxes have clear lids with locator grid to facilitate sample identification. Polycarbonate boxes are designed to withstand temperature extremes of 196 to 121 . Boxes have drain holes for venting and safety. Keyed lid prevents misalignment of grid. Features: Fit all the most popular cryo and cold storage vials. Fit into standard freezer racks. Boxes have clear lids with locator grid to facilitate sample identification. Withstand temperature extremes of 196°C to 121°C. Drain holes for venting and safety. Keyed lid prevents misalignment of grid.