Learn More
Chemglass Life Sciences 2-Port Assembly for 6L VSA Vessels and 5L Media Bottles, Complete
SDP
Catalog No. 50143180
Encompass
Our probe holders are designed to secure pH and DO probes into 45mm vertical sidearm vessels.Includes thermoplastic, polyester cap and silicone gasket.This assembly is designed to hold an Ingold pH or DO electrode securely in place in your cell culture vessel via the 45mm sidearm.It consists of: red high temperature 45mm screw cap, 316L SST probe holder with a Pg 13.5 thread, and a white silicone rubber liner. comes fully assembled for fast and easy set-up.
Catalog No. 50-143-180 Supplier Chemglass Life Sciences Supplier No. CLS143721
Our probe holders are designed to secure pH and DO probes into 45mm vertical sidearm vessels.Includes thermoplastic, polyester cap and silicone gasket.This assembly is designed to hold an Ingold pH or DO electrode securely in place in your cell culture vessel via the 45mm sidearm.It consists of: red high temperature 45mm screw cap, 316L SST probe holder with a Pg 13.5 thread, and a white silicone rubber liner. comes fully assembled for fast and easy set-up.