Learn More
Chemglass Life Sciences 2-Port Drip Tube Assembly with Open Top Cap and Gasket
SDP
Catalog No. 50143209
Encompass
2-Port Drip Tube Assembly with Open Top Cap and Gasket. Replacement part for item 50143731. Assembly is ideal for use with bio reactors where addition or sampling is required. The drip tube assemblies are constructed of 316L electropolished stainless steel and include a blue polypropylene GL32 open top cap with a white silicone gasket. Drip tubes have a finish of 20Ra or better. Our addition and sampling vessel with hose barbed 2-port drip tube assemblies are designed for addition or sampling of media, acid or base.
Catalog No. 50-143-209 Supplier Chemglass Life Sciences Supplier No. CLS142902
2-Port Drip Tube Assembly with Open Top Cap and Gasket. Replacement part for item 50143731. Assembly is ideal for use with bio reactors where addition or sampling is required. The drip tube assemblies are constructed of 316L electropolished stainless steel and include a blue polypropylene GL32 open top cap with a white silicone gasket. Drip tubes have a finish of 20Ra or better. Our addition and sampling vessel with hose barbed 2-port drip tube assemblies are designed for addition or sampling of media, acid or base.