Learn More
Bellco Glass, Inc. 2-Port Sampler Assembly , 32mm For Larger 32mm Side Arms's
SDP
Catalog No. 501945789
Encompass_Preferred
Universal port assembly for 32mm sidearms2 -port sampler assembly, 316l sst materialBoth tubes: .187"od x .117" idSupplied with 32mm s/cap & silicone linerThis port cap assembly can be used on any usage of 32mm sidearms. MADE IN THE USA
Catalog No. 50-194-5789 Supplier Bellco Glass, Inc. Supplier No. 563751000
Universal port assembly for 32mm sidearms2 -port sampler assembly, 316l sst materialBoth tubes: .187"od x .117" idSupplied with 32mm s/cap & silicone linerThis port cap assembly can be used on any usage of 32mm sidearms. MADE IN THE USA