Learn More
Chemglass Life Sciences 2-Port Angled Vent Assembly, GL-32, Universal, 316L Electro Polished Stainless Steel, 3/16" OD Hose Barbs, Complete with Gasket and Cap
SDP
Catalog No. 501214877
Encompass
Universal vent port assembly is constructed of 316L electro polished stainless steel.These port assemblies fit all types of spinner flasks, media bottles and vessels.Configurations consist of a single vent tube, vent and short fill tubes, or angled vent and fill tubes.Supplied complete with a GL-32 or GL-45 open top cap and silicone gasket.
Catalog No. 50-121-4877 Supplier Chemglass Life Sciences Supplier No. CLS142901
Universal vent port assembly is constructed of 316L electro polished stainless steel.These port assemblies fit all types of spinner flasks, media bottles and vessels.Configurations consist of a single vent tube, vent and short fill tubes, or angled vent and fill tubes.Supplied complete with a GL-32 or GL-45 open top cap and silicone gasket.