Learn More
Bellco Glass, Inc. 2-Port Stainless Steel Assembly , 8L For Vertical Over Head Drive Vessels
SDP
Catalog No. 501944778
Encompass_Preferred
2-port 45mm cap assembly, 8lFor 8l 1964 style Over Head Drive vessels. MADE IN THE USALong tube: 1/4"od x 16 1/4"Short tube: 1/4"od x 2.5/32"All tubes extended up 1.0" above the cap slugSupply 45mm s/cap & white silicone liner
Catalog No. 50-194-4778 Supplier Bellco Glass, Inc. Supplier No. 196420008R
2-port 45mm cap assembly, 8lFor 8l 1964 style Over Head Drive vessels. MADE IN THE USALong tube: 1/4"od x 16 1/4"Short tube: 1/4"od x 2.5/32"All tubes extended up 1.0" above the cap slugSupply 45mm s/cap & white silicone liner