Learn More
MTC Bio DuoClick™ Screw-Cap Culture Tubes
Catalog No. NC1819959
Encompass
Culture Tube, 14mL, 17 x 100mm, PP, w/ separate 2-position screw-cap, non-sterile, non-graduated, 1000 tubes and capped bulk packed separately
Catalog No. NC1819959 Supplier MTC Bio Supplier No. T8850
Culture Tube, 14mL, 17 x 100mm, PP, w/ separate 2-position screw-cap, non-sterile, non-graduated, 1000 tubes and capped bulk packed separately