Learn More
MTC Bio DuoClick™ Screw-Cap Culture Tubes
Supplier:  MTC Bio T8832
Encompass
Culture Tube, 14mL, 17 x 100mm, PP, w/ attached 2-position screw-cap, printed graduations, sterile, 10 foam racks of 50 tubes, 500/case
Catalog No. NC1819957