Learn More
MTC Bio DuoClick™ Screw-Cap Culture Tubes
Supplier:  MTC Bio T8730
Encompass
Culture Tube, 5mL, 12 x 75mm, PP, w/ attached 2-position screw-cap, printed graduations, sterile, 20 bags of 25 tubes, 500/case
Catalog No. NC1819952