Learn More
Waters Corp 2-Position Regeneration Valve Upgrade, 10-Port
Catalog No. 50791620 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This upgrade kit provides essential components for a 2-position regeneration valve, designed to enhance the performance and longevity of your analytical system. It ensures efficient and reliable operation for various laboratory applications.
  • Kit includes a 2-position regeneration valve with 10 ports.
  • PEEK tubing (0.062 in. x 0.010 in., 300 in. long) for connections.
  • Compression screws for secure connections.
  • Valve liquid contacts constructed from stainless steel and alumina ceramic.
  • Maximum pressure rating of 35 MPa (5000 psi).
  • Supports automated operation and remote control.
  • Compatible with a wide range of laboratory computer systems.
Catalog No. 50-791-620 Supplier Waters Corp Supplier No. 205000122
Add to Cart
Add to Cart
Request a Quote
This upgrade kit provides essential components for a 2-position regeneration valve, designed to enhance the performance and longevity of your analytical system. It ensures efficient and reliable operation for various laboratory applications.
  • Kit includes a 2-position regeneration valve with 10 ports.
  • PEEK tubing (0.062 in. x 0.010 in., 300 in. long) for connections.
  • Compression screws for secure connections.
  • Valve liquid contacts constructed from stainless steel and alumina ceramic.
  • Maximum pressure rating of 35 MPa (5000 psi).
  • Supports automated operation and remote control.
  • Compatible with a wide range of laboratory computer systems.