Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Propanesulfonyl chloride

Catalog No. 24270525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-270-525G 25 g
24-270-5100G 100 g
24-270-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 24-270-525G Supplier Sigma Aldrich Supplier No. 24270525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The solvolysis of 2-propanesulfonyl chloride by an addition-elimination (association-dissociation) pathway was studied.2-propanesulfonyl chloride (isopropylsulfonyl chloride) can be used as a reactant to prepare:
  • 1-isopropylsulfonyl-2-amine benzimidazole by reacting with 2-aminobenzimidazole via n-sulfonylation reaction in the presence of a base
  • Bis(isopropylsulfonyl)disulfide, a sulfurizing agent, used to synthesize phosphorothioate oligonucleotide analogs

Specifications

CAS 10147-37-2
Density 1.27 g/mL (at 25°C (literature))
Boiling Point 74°C to 75°C (19 mmHg)
Molecular Formula C3H7ClO2S
Refractive Index n20/D 1.453 (literature)
Linear Formula (CH3)2 CHSO2Cl
MDL Number MFCD00007453
Quantity 25 g
Molecular Weight (g/mol) 142.6
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.