Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Propanethiol, 98%
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
2-Propanethiol Reacts with 3-chloro-propene to get 3-isopropylsulfanyl-propene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
| Melting Point | -131°C |
| Color | Colorless |
| Density | 0.82 g/mL |
| Boiling Point | 57°C to 60°C |
| Flash Point | -34° (-29°F) |
| Odor | Stench |
| Refractive Index | 1.4255 |
| Linear Formula | (CH3)2CHSH |
| Quantity | 25 mL |
| UN Number | UN2402 |
| Show More |
Safety and Handling
| Hazard Category | H225-H317 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P272-P280-P303+P361+P353-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPANETHIOLS |
| EINECSNumber | 200-861-4 |
| RTECSNumber | TZ7302000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction