Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propanol, 1,3-dibromo- | 96-21-9 | 99.7% | 217.89 g/mol | C3H6Br2O | 10 G
SDP

Catalog No. 5000303818
Encompass_Preferred

1,3-Dibromo-2-propanol is an organic biochemical reagent used as a building block and intermediate in chemical synthesis and research applications. It is provided as a high-purity liquid for laboratory use and should be handled according to applicable safety data and institutional protocols.

  • High purity (99.72%) suitable for analytical and synthesis work.
  • Molecular weight 217.89 g/mol; formula C3H6Br2O.
  • Colorless to light yellow liquid with consistent quality.
  • Available in multiple pack sizes, including small-scale quantities for research.
  • Storage: stable at room temperature for up to 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Supplied for research use only; not for human or clinical use.

Catalog No. 50-003-03818 Supplier Medchemexpress LLC Supplier No. HYW01409210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dibromo-2-propanol is an organic biochemical reagent used as a building block and intermediate in chemical synthesis and research applications. It is provided as a high-purity liquid for laboratory use and should be handled according to applicable safety data and institutional protocols.

  • High purity (99.72%) suitable for analytical and synthesis work.
  • Molecular weight 217.89 g/mol; formula C3H6Br2O.
  • Colorless to light yellow liquid with consistent quality.
  • Available in multiple pack sizes, including small-scale quantities for research.
  • Storage: stable at room temperature for up to 3 years; in solvent: -80°C for 2 years, -20°C for 1 year.
  • Supplied for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.