Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propanol, 1,3-dibromo- | 96-21-9 | MFCD00000216 | 99.7% | 217.89 g·mol⁻¹ | C3H6Br2O | 1 KG
SDP

Catalog No. 5000304002
Encompass_Preferred

1,3-Dibromo-2-propanol is a dihalogenated propanol used as an intermediate and reagent in organic synthesis and biochemical research. It is supplied with a certificate of analysis for identity and purity; consult the SDS and COA for handling, storage, and batch-specific information.

  • molecular formula: C3H6Br2O
  • molecular weight: 217.89 g·mol⁻¹
  • appearance: colorless to light yellow liquid
  • purity (GC): 99.72%
  • applications: reagent for organic synthesis and biochemical assays
  • packaging: available in laboratory-scale and bulk quantities

Catalog No. 50-003-04002 Supplier Medchemexpress LLC Supplier No. HYW0140921KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3-Dibromo-2-propanol is a dihalogenated propanol used as an intermediate and reagent in organic synthesis and biochemical research. It is supplied with a certificate of analysis for identity and purity; consult the SDS and COA for handling, storage, and batch-specific information.

  • molecular formula: C3H6Br2O
  • molecular weight: 217.89 g·mol⁻¹
  • appearance: colorless to light yellow liquid
  • purity (GC): 99.72%
  • applications: reagent for organic synthesis and biochemical assays
  • packaging: available in laboratory-scale and bulk quantities

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.