Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propanol, 1,3-dibromo- | 96-21-9 | MFCD00000216 | 99.7% | 217.89 g/mol | C3H6Br2O | 25 G
SDP

Catalog No. 5000303007
Encompass_Preferred

1,3-Dibromo-2-propanol is a dibromo-substituted secondary alcohol used as a synthetic intermediate and biochemical reagent in laboratory research. It is supplied as a colorless to light yellow liquid with high stated purity and is suitable for organic synthesis and analytical applications.

  • High purity (99.72%).
  • Molecular formula C3H6Br2O; molecular weight 217.89 g/mol.
  • Available in multiple pack sizes for scale flexibility.
  • Storage: room temperature shelf life; in solvent long-term storage at -80°C or -20°C as specified.
  • Colorless to light yellow liquid, convenient for handling.

Catalog No. 50-003-03007 Supplier Medchemexpress LLC Supplier No. 25GHYW014092
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1,3-Dibromo-2-propanol is a dibromo-substituted secondary alcohol used as a synthetic intermediate and biochemical reagent in laboratory research. It is supplied as a colorless to light yellow liquid with high stated purity and is suitable for organic synthesis and analytical applications.

  • High purity (99.72%).
  • Molecular formula C3H6Br2O; molecular weight 217.89 g/mol.
  • Available in multiple pack sizes for scale flexibility.
  • Storage: room temperature shelf life; in solvent long-term storage at -80°C or -20°C as specified.
  • Colorless to light yellow liquid, convenient for handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.