Learn More
Medchemexpress LLC 2-Propanol, 1-[4-(2-methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-, (2R,3R)-2,3-dihydroxybutanedioate (2:1) | 56392-17-7 | 99.98% | 342.41 | 25MG
SDP
Catalog No. 5000384117
Encompass_Preferred
Metoprolol tartrate is characterized as an orally active, selective β1-adrenoceptor antagonist. It has demonstrated various biological activities, including anti-inflammatory, antitumor, and anti-angiogenic properties. Studies have shown its ability to reduce proinflammatory cytokines and atherosclerosis, protect myocardial tissue from viral damage, decrease myocardial virus titers, and inhibit myocardial apoptosis.
  • Orally active compound.
  • Selective antagonist of a specific adrenoceptor.
  • Exhibits anti-inflammatory effects.
  • Demonstrates antitumor properties.
  • Possesses anti-angiogenic activity.
  • Reduces proinflammatory cytokines.
  • Decreases atherosclerosis.
  • Protects heart muscle from viral damage.
  • Lowers myocardial virus titers.
  • Inhibits programmed cell death in heart tissue.
Catalog No. 50-003-84117 Supplier Medchemexpress LLC Supplier No. 25MGHY17503B
Metoprolol tartrate is characterized as an orally active, selective β1-adrenoceptor antagonist. It has demonstrated various biological activities, including anti-inflammatory, antitumor, and anti-angiogenic properties. Studies have shown its ability to reduce proinflammatory cytokines and atherosclerosis, protect myocardial tissue from viral damage, decrease myocardial virus titers, and inhibit myocardial apoptosis.
  • Orally active compound.
  • Selective antagonist of a specific adrenoceptor.
  • Exhibits anti-inflammatory effects.
  • Demonstrates antitumor properties.
  • Possesses anti-angiogenic activity.
  • Reduces proinflammatory cytokines.
  • Decreases atherosclerosis.
  • Protects heart muscle from viral damage.
  • Lowers myocardial virus titers.
  • Inhibits programmed cell death in heart tissue.