Learn More
U.S. Pharmacopeia 2-Propanol, 67-63-0, MFCD00011674, 1.2 mL/ampule; 3 ampules
Catalog No. NC9871848
Encompass
Molecular Formula C3H8O, Molecular Weight 60.095, Melting Point -127.3 °F (-88.5 °C), Boiling Point 180.5 °F (82.5 °C) 101.325 kPa, Flash Point 53.6 °F (12.0 °C) Closed Cup, 75.0 °F (23.9 °C) Open Cup, Synonyms: 1-Methylethanol * Isopropanol
Catalog No. NC9871848 Supplier U.S. Pharmacopeia Supplier No. 1570428
Molecular Formula C3H8O, Molecular Weight 60.095, Melting Point -127.3 °F (-88.5 °C), Boiling Point 180.5 °F (82.5 °C) 101.325 kPa, Flash Point 53.6 °F (12.0 °C) Closed Cup, 75.0 °F (23.9 °C) Open Cup, Synonyms: 1-Methylethanol * Isopropanol