Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Propanol, 67-63-0, MFCD00011674, 500mL
Catalog No. 501657113
Encompass
Linear Formula (CH3)2CHOH, Purity ≥99.5%, Molecular Weight 60.10, BioReagent, for molecular biology, Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
Catalog No. 50-165-7113 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I9516500ML
Linear Formula (CH3)2CHOH, Purity ≥99.5%, Molecular Weight 60.10, BioReagent, for molecular biology, Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol