Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Propanol, 67-63-0, MFCD00011674, 500 mL
Catalog No. NC0889287
Encompass
Linear Formula: (CH3)2CHOH, Molecular Weight: 60.10, BioUltra, for molecular biology, ≥99.5% (GC), Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol. Isopropanol (2-Propanol) is widely used in molecular biology for the precipitation of DNA its during extraction from cells and tissues. BioUltra IPA is suitable for DNA precipitation.
Catalog No. NC0889287 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 59304500MLF
Add to Cart
Add to Cart
Linear Formula: (CH3)2CHOH, Molecular Weight: 60.10, BioUltra, for molecular biology, ≥99.5% (GC), Synonym: sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol. Isopropanol (2-Propanol) is widely used in molecular biology for the precipitation of DNA its during extraction from cells and tissues. BioUltra IPA is suitable for DNA precipitation.