Learn More
2-Propanol, ACS, 99.5% min
Description
It acts as a solvent for gums, resins, alkaloids and essential oils. It is used in cosmetic lotions. Isopropyl alcohol is used as a cleansing agent for keyboards, LCD displays and laptops. It is used as a wetting agent in the fountain solution used in lithographic printing, and is often used as a solvent for french polishing shellac, which is used in cabinet making. It act as a hydride source in the Meerwein-Ponndorf-Verley reduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 90°C |
| Color | Colorless |
| Appearance | Liquid |
| Density | 0.785 g/cm3 at 20°C |
| Quantity | 1 L |
| Flash Point | 12°C (53°F) |
| Odor | Alcohol-like |
| Physical Form | Liquid |
| Refractive Index | 1.377 |
| Assay Percent Range | 99.5% |
| Show More |
Safety and Handling
| Hazard Category | H225-H319-H335-H336-H373 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c |
| DOTInformation | DOT Class: 3; Packing Group: II |
| EINECSNumber | 200-661-7 |
| RTECSNumber | NT8050000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.