Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, ACS, 99.5% min

Catalog No. AA36644M6
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36644M6 4 x 4 L
AA36644K2 1 L
AA36644K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36644M6 Supplier Thermo Scientific Chemicals Supplier No. 036644M6

missing translation for 'validMainframeMsg'

Solvent For Gums, Resins, Alkaloids | CAS: 67-63-0 | C3H8O | 60.096 g/mol

It acts as a solvent for gums, resins, alkaloids and essential oils. It is used in cosmetic lotions. Isopropyl alcohol is used as a cleansing agent for keyboards, LCD displays and laptops. It is used as a wetting agent in the fountain solution used in lithographic printing, and is often used as a solvent for french polishing shellac, which is used in cabinet making. It act as a hydride source in the Meerwein-Ponndorf-Verley reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point °C to 90°C
Color Colorless
Appearance Liquid
Density 0.785
Quantity 4 x 4 L
Flash Point 12°C (53°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.377
Assay Percent Range 99.5%
Beilstein 635639
Solubility Information Miscible with water,alcohol,ether,chloroform,acetone and benzene.
Boiling Point 82°C to 83°C
Formula Weight 60.1
Grade ACS
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Category H225-H319-H335-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: II
EINECSNumber 200-661-7
RTECSNumber NT8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.