Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Propanol ACS reagent 99.
Catalog No. 502897827
Encompass
2-Propanol is a secondary alcohol. It has been tested as a substitute fuel for use in various fuel cells. CuO powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Suspension of 2-propanol with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with a uniformly thick layer of P25-TiO2. This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process.
Catalog No. 50-289-7827 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1907642.5L
2-Propanol is a secondary alcohol. It has been tested as a substitute fuel for use in various fuel cells. CuO powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Suspension of 2-propanol with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with a uniformly thick layer of P25-TiO2. This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process.