Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol (Certified ACS Plus), Fisher Chemical

Catalog No. A416SK4001 Shop All Fisherbrand Products
Change view
Click to view available options
4L; Safe-Cote
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A416SK4-001 4L; Safe-Cote
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A416SK4-001

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Specifications

pH 7
Melting Point -89.5°C
CAS 67-63-0
Color Colorless
Density 0.785g/cm³
Quantity 4 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Packaging Safe-Cote™
Physical Form Liquid
Refractive Index 1.3740 to 1.3760 (at 25°C)
Assay Percent Range ≥99.5 %
Residue after Evaporation 0.001% max.
Specific Gravity 0.785
Molecular Formula C3H8O
Linear Formula (CH3)2CHOH
MDL Number MFCD00011674
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPa/s at 20°C
UN Number 1219
Water 0.2% max.
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Boiling Point 81°C
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material 2-Propanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipm
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.