Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol CHROMASOLV™, for HPLC, 99.9%, Solstice

Catalog No. 6000129 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
185 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-001-29 4 x 4 L
60-001-23 1 L
60-001-28 4 x 2.5 L
60-001-22 185 L
60-001-24 2.5 L
60-001-30 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-001-29 Supplier Solstice Supplier No. 348634X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for HPLC, 99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Density 0.785g/mL at 25°C
Quantity 4 x 4 L
Refractive Index n20/D 1.377
Vapor Pressure 44mmHg (25°C)
Linear Formula (CH3)2CHOH
Beilstein 635639
Boiling Point 82°C
Formula Weight 60.1g/mol
Percent Purity 99.9%
Grade CHROMASOLV
Chemical Name or Material 2-Propanol, for HPLC
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
EINECSNumber 200-661-7
RTECSNumber NT8050000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.