Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol-d{8}, 99%(Isotopic)

Catalog No. AA4231414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4231414 25 g
AA4231406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4231414 Supplier Thermo Scientific Chemicals Supplier No. 4231414
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 22739-76-0 | C3H8O | 68.15 g/mol

2-Propanol-d{8} is used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22739-76-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 68.15
MDL Number MFCD00044341
InChI Key KFZMGEQAYNKOFK-PIODKIDGSA-N
Synonym 2-propanol-d8, isopropanol-d8, 2 h? propan-2-2 h ol, octadeuteroisopropanol, 2h8-2-propanol, 2-propan-1,1,1,2,3,3,3-d7-ol-d, 2-propanol-d isotopic 1g, 2-propanol-d8, 99.5 atom % d
PubChem CID 2723972
SMILES [2H]OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Quantity 25 g
Density 0.90 g/mL
Flash Point −11.7°C (53°F)
Odor Alcohol-like
Melting Point -89.5°C
Boiling Point 82.4°C
UN Number UN1219
Solubility Information Completely soluble in water.
IUPAC Name 1,1,1,2,3,3,3-heptadeuterio-2-deuteriooxypropane
Formula Weight 68.15
Percent Purity 99%
Chemical Name or Material 2-Propanol-d8, Isotopic
Show More Show Less
Hazard Category H225-H319-H336
Hazard Statement GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
EINECSNumber 245-189-2
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.