Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, HPLC, J.T. Baker™

Catalog No. 14650208 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Glass Bottle
Poly Container
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
14-650-208 1 L Poly Container
02-002-094 4 L Narrow Mouth Amber Glass Bottle
02-002-095 4 L Narrow Mouth PVC-coated Amber Glass Bottle
14-650-220 4 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-650-208 Supplier Avantor Supplier No. 909502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Quantity 1 L
Packaging Poly Container
Formula Weight 60.1
Percent Purity ≥99.7%
Grade HPLC
Chemical Name or Material 2-Propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.