Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™

Catalog No. 01100234 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-100-234 2.5 L
01-100-235 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-100-234 Supplier MilliporeSigma Supplier No. 1.02781.2500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.1
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropyl alcohol, Isopropanol, iso-Propanol, IPA
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Density 0.78 g/cm3 at 20 °C
Quantity 2.5 L
Explosion Limit 2 to 13.4% (V)
Flash Point 12 °C
Ignition Point 425 °C
Packaging Glass Bottle
Residue after Evaporation ≤ 2 mg/L
Vapor Pressure 43 hPa at 20 deg;C
Water ≤ 0.05%
Boiling Point 82.4°C (1013 hPa)
Percent Purity ≥99.9%
Grade Hypergrade
Show More Show Less
Recommended Storage Store at 2 to 30 °C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.