Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, puriss., ≥99.5% (GC), Solstice

Catalog No. 60045537 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
160 kg
Packaging:
Metal Drum
Poly Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-537 6 x 1 L Poly Bottle
60-045-530 1 L Poly Bottle
60-045-536 5 L Poly Bottle
60-045-534 4 x 2.5 L Poly Bottle
60-045-532 20 kg Metal Drum
60-045-535 4 x 5 L Poly Bottle
60-045-529 160 kg Metal Drum
60-045-531 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 60-045-537 Supplier Solstice Supplier No. 241376X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of BP, Ph. Eur., USP,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -85°C
Color Colorless
Density 0.785g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Poly Bottle
Physical Form Liquid
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
Linear Formula (CH3)2CHOH
UN Number UN1219
Beilstein 635639
Boiling Point 82°C
Formula Weight 60.1g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material 2-Propanol, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
DOTInformation class 3 - PG 2 - Isopropanol
EINECSNumber 200-661-7
RTECSNumber NT8050000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.