Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Propanol suitable for HP
Catalog No. 502906626
Encompass
2-Propanol (Isopropanol) is a secondary alcohol. It has been tested as substitute to fuel for various fuel cells. Cuo powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Its suspension with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with uniformly thick layer of P25-TiO2. This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process.
Catalog No. 50-290-6626 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348632.5L
Add to Cart
Add to Cart
2-Propanol (Isopropanol) is a secondary alcohol. It has been tested as substitute to fuel for various fuel cells. Cuo powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Its suspension with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with uniformly thick layer of P25-TiO2. This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process.