Learn More
Medchemexpress LLC 2-Propanone, 1-hydroxy-3-(phosphonooxy)- | 57-04-5 | 99.9% | 170.06 | 100MG
SDP
Catalog No. 5000435754
Encompass_Preferred
Dihydroxyacetone phosphate (DHPA), a derivative of Dihydroxyacetone (DHA), is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research.
  • Participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle.
  • Can be used as a substrate and metabolic marker in biochemical research.
  • Relevant to endogenous metabolite research.
  • Useful as a biochemical assay reagent.
  • Important in metabolic disease research.
Catalog No. 50-004-35754 Supplier Medchemexpress LLC Supplier No. HY113131100MG
Dihydroxyacetone phosphate (DHPA), a derivative of Dihydroxyacetone (DHA), is an important intermediate that participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle. It can be used as a substrate and metabolic marker in biochemical research.
  • Participates in key pathways including glycolysis, lipid biosynthesis, and the plant Calvin cycle.
  • Can be used as a substrate and metabolic marker in biochemical research.
  • Relevant to endogenous metabolite research.
  • Useful as a biochemical assay reagent.
  • Important in metabolic disease research.