Learn More
Medchemexpress LLC 2-propen-1-one, 1,3-diphenyl- | 94-41-7 | 100.0% | 1 ML
SDP
Catalog No. 5000232558
Encompass_Preferred
Chalcone (2-Propen-1-one, 1,3-diphenyl-) is an α,β-unsaturated ketone used as a synthetic building block and research reagent. It is supplied as a solid or as a prepared 10 mM solution in DMSO, 1 ML, for screening and synthesis applications.
  • Offered as solid and as 10 mM solution in DMSO, 1 ML.
  • High purity (99.98% by HPLC) suitable for research use.
  • Commonly used for synthesis of chalcone derivatives and biological screening.
  • Provided with certificate of analysis documenting identity and purity.
Catalog No. 50-002-32558 Supplier Medchemexpress LLC Supplier No. HY12105410MM 1ML
Chalcone (2-Propen-1-one, 1,3-diphenyl-) is an α,β-unsaturated ketone used as a synthetic building block and research reagent. It is supplied as a solid or as a prepared 10 mM solution in DMSO, 1 ML, for screening and synthesis applications.
  • Offered as solid and as 10 mM solution in DMSO, 1 ML.
  • High purity (99.98% by HPLC) suitable for research use.
  • Commonly used for synthesis of chalcone derivatives and biological screening.
  • Provided with certificate of analysis documenting identity and purity.