Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propen-1-one, 1,3-diphenyl- | 94-41-7 | 100.0% | 100 MG
SDP

Catalog No. 5000231976
Encompass_Preferred

Chalcone is a small-molecule research chemical used as a synthetic intermediate and a reference compound in medicinal chemistry and biological assays. Offered in solid and solution formats, it is supplied with a certificate of analysis for batch verification.

  • High purity suitable for analytical and synthetic work.
  • Off-white to light yellow solid for easy handling.
  • Soluble in DMSO at up to 100 mg/mL with sonication.
  • Recommended storage for powder at -20°C for long-term stability.
  • Certificate of analysis available for batch verification.

Catalog No. 50-002-31976 Supplier Medchemexpress LLC Supplier No. HY121054100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chalcone is a small-molecule research chemical used as a synthetic intermediate and a reference compound in medicinal chemistry and biological assays. Offered in solid and solution formats, it is supplied with a certificate of analysis for batch verification.

  • High purity suitable for analytical and synthetic work.
  • Off-white to light yellow solid for easy handling.
  • Soluble in DMSO at up to 100 mg/mL with sonication.
  • Recommended storage for powder at -20°C for long-term stability.
  • Certificate of analysis available for batch verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.