Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propen-1-one, 3-(3-pyridinyl)-1-(4-pyridinyl)- | 13309-08-5 | 99.9% | 210.24 | 25 MG
SDP

Catalog No. 5000345867
Encompass_Preferred

The (E/Z)-3PO compound, with chemical formula C13H10N2O and CAS number 13309-08-5, is intended for laboratory research and for use in the manufacture of substances. It is an off-white to light yellow solid, stable under recommended storage conditions, and exhibits a high purity of 99.9% by LCMS. The compound should be handled by experienced personnel in appropriately equipped facilities for research use only.

  • Identified for laboratory research applications
  • Suitable for substance manufacturing
  • Solid appearance, off-white to light yellow
  • Melting point between 144-147 °C
  • Boiling point range of 387.8±42.0 °C at 760 mmHg
  • Stable under recommended storage conditions
  • Molecular weight of 210.24

Catalog No. 50-003-45867 Supplier Medchemexpress LLC Supplier No. 25MGHY110169
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The (E/Z)-3PO compound, with chemical formula C13H10N2O and CAS number 13309-08-5, is intended for laboratory research and for use in the manufacture of substances. It is an off-white to light yellow solid, stable under recommended storage conditions, and exhibits a high purity of 99.9% by LCMS. The compound should be handled by experienced personnel in appropriately equipped facilities for research use only.

  • Identified for laboratory research applications
  • Suitable for substance manufacturing
  • Solid appearance, off-white to light yellow
  • Melting point between 144-147 °C
  • Boiling point range of 387.8±42.0 °C at 760 mmHg
  • Stable under recommended storage conditions
  • Molecular weight of 210.24

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.