Learn More
Medchemexpress LLC 2-propenal, 3-[4-(dimethylamino)phenyl]- | 6203-18-5 | MFCD00007002 | >97.5% | 175.23 g/mol | C11H13NO | 5 G
SDP
Catalog No. 5000299389
Encompass_Preferred
4-(Dimethylamino)cinnamaldehyde (CAS 6203-18-5) is a small-molecule chromogenic reagent used to detect indole-containing compounds and flavanols in biochemical assays. Supplied as a solid for research use, it dissolves readily in DMSO to prepare concentrated stock solutions for assay development.
  • Chromogenic reagent for indoles and flavanols.
  • Dissolves in DMSO at 100 mg/mL with ultrasonic warming to 60°C.
  • Molecular weight 175.23 g/mol; chemical formula C11H13NO.
  • Powder storage recommended at 4°C (2 years) or -20°C (3 years); in solution store at -20°C (1 month) or -80°C (6 months).
  • Supplied in research-scale quantities suitable for preparing mg-scale stock solutions.
Catalog No. 50-002-99389 Supplier Medchemexpress LLC Supplier No. HYW0167295G
4-(Dimethylamino)cinnamaldehyde (CAS 6203-18-5) is a small-molecule chromogenic reagent used to detect indole-containing compounds and flavanols in biochemical assays. Supplied as a solid for research use, it dissolves readily in DMSO to prepare concentrated stock solutions for assay development.
  • Chromogenic reagent for indoles and flavanols.
  • Dissolves in DMSO at 100 mg/mL with ultrasonic warming to 60°C.
  • Molecular weight 175.23 g/mol; chemical formula C11H13NO.
  • Powder storage recommended at 4°C (2 years) or -20°C (3 years); in solution store at -20°C (1 month) or -80°C (6 months).
  • Supplied in research-scale quantities suitable for preparing mg-scale stock solutions.