Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-propenamide, 3-(3-chlorophenyl)-N-[2-[4-(methylsulfonyl)-1-piperazinyl]-2-oxoethyl]-, (2E)- | 2505001-54-5 | 100.0% | 385.87 g·mol-1 | C16H20ClN3O4S | 5 MG
SDP

Catalog No. 5000221429
Encompass_Preferred

SR15006 is a small-molecule inhibitor of Krüppel-like factor 5 (KLF5) supplied as a high-purity research reagent. It has reported activity in cell-based assays (IC50 = 41.6 nM in DLD-1/pGL4.18hKLF5p cells) and is provided with solubility and storage guidance for in vitro and in vivo use.

  • Inhibits KLF5 with IC50 = 41.6 nM in DLD-1/pGL4.18hKLF5p cells.
  • High purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL; example in vivo formulations provided.
  • Recommended storage: solid at 4°C; in solution -80°C (6 months) or -20°C (1 month).
  • Molecular formula C16H20ClN3O4S; molecular weight 385.87 g·mol-1.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-21429 Supplier Medchemexpress LLC Supplier No. HY1396915MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SR15006 is a small-molecule inhibitor of Krüppel-like factor 5 (KLF5) supplied as a high-purity research reagent. It has reported activity in cell-based assays (IC50 = 41.6 nM in DLD-1/pGL4.18hKLF5p cells) and is provided with solubility and storage guidance for in vitro and in vivo use.

  • Inhibits KLF5 with IC50 = 41.6 nM in DLD-1/pGL4.18hKLF5p cells.
  • High purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL; example in vivo formulations provided.
  • Recommended storage: solid at 4°C; in solution -80°C (6 months) or -20°C (1 month).
  • Molecular formula C16H20ClN3O4S; molecular weight 385.87 g·mol-1.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.