Learn More
Medchemexpress LLC 2-Propenamide, 3-(3-chlorophenyl)-N-[2-[4-(methylsulfonyl)-1-piperazinyl]-2-oxoethyl]-, (2E)- | 2505001-54-5 | 99.9% | 385.87 | 50 MG
SDP
Catalog No. 5000350917
Encompass_Preferred
SR15006 is an inhibitor of Krüppel-like factor 5 (KLF5) with an IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells. It causes a significant reduction in the levels of tested cyclins over 72 hours in DLD-1 CRC cells at concentrations of 1 μM or 10 μM. This product is for research use only.
  • Inhibits Krüppel-like factor 5 (KLF5)
  • IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells
  • Causes a significant reduction in cyclin levels over 72 hours
  • Appearance: solid, white to off-white
Catalog No. 50-003-50917 Supplier Medchemexpress LLC Supplier No. HY13969150MG
SR15006 is an inhibitor of Krüppel-like factor 5 (KLF5) with an IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells. It causes a significant reduction in the levels of tested cyclins over 72 hours in DLD-1 CRC cells at concentrations of 1 μM or 10 μM. This product is for research use only.
  • Inhibits Krüppel-like factor 5 (KLF5)
  • IC50 of 41.6 nM in DLD-1/pGL4.18hKLF5p cells
  • Causes a significant reduction in cyclin levels over 72 hours
  • Appearance: solid, white to off-white